accessibility menu, dialog, popup

UserName

NBrBrCl

CAS RN 137628-17-2

IUPAC Name: 2,3-dibromo-5-chloropyridine
Synonyms: 2,3-dibromo-5-chloropyridine
Molecular Weight (g/mol): 271.34
Molecular Formula: C5H2Br2ClN
InChi Key: GDUFWKJMOOVEMX-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=C(Br)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results