missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Diaminonaphthalene 98.0+%, TCI America™
$247.92 - $555.49
Chemical Identifiers
| CAS | 771-97-1 |
|---|---|
| Molecular Formula | C10H10N2 |
| Molecular Weight (g/mol) | 158.204 |
| MDL Number | MFCD00004116 |
| InChI Key | XTBLDMQMUSHDEN-UHFFFAOYSA-N |
| Synonym | 2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532 |
| PubChem CID | 69872 |
| IUPAC Name | naphthalene-2,3-diamine |
| SMILES | C1=CC=C2C=C(C(=CC2=C1)N)N |
Chemical Identifiers
| 771-97-1 | |
| 158.204 | |
| XTBLDMQMUSHDEN-UHFFFAOYSA-N | |
| 69872 | |
| C1=CC=C2C=C(C(=CC2=C1)N)N |
| C10H10N2 | |
| MFCD00004116 | |
| 2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532 | |
| naphthalene-2,3-diamine |
Specifications
| 771-97-1 | |
| C10H10N2 | |
| 1 g | |
| XTBLDMQMUSHDEN-UHFFFAOYSA-N | |
| naphthalene-2,3-diamine | |
| 69872 | |
| ≥98.0% (T) | |
| 2,3-Diaminonaphthalene |
| 198°C | |
| MFCD00004116 | |
| 2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532 | |
| C1=CC=C2C=C(C(=CC2=C1)N)N | |
| 158.204 | |
| 158.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator