missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Diaminonaphthalene 98.0+%, TCI America™
Supplier: TCI America D10455G
Specifications
| 2,3-Diaminonaphthalene | |
| 198°C | |
| MFCD00004116 | |
| 2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532 | |
| C1=CC=C2C=C(C(=CC2=C1)N)N | |
| 158.204 | |
| 158.20 | |
| Crystalline Powder |
| 771-97-1 | |
| C10H10N2 | |
| 5 g | |
| XTBLDMQMUSHDEN-UHFFFAOYSA-N | |
| naphthalene-2,3-diamine | |
| 69872 | |
| ≥98.0% (T) |
Chemical Identifiers
| 771-97-1 | |
| 158.204 | |
| XTBLDMQMUSHDEN-UHFFFAOYSA-N | |
| 69872 | |
| C1=CC=C2C=C(C(=CC2=C1)N)N |
| C10H10N2 | |
| MFCD00004116 | |
| 2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532 | |
| naphthalene-2,3-diamine |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator