missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminobenzoic acid, 95%

Catalog No. AC457490010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457490010 1 g
AC457490050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457490010 Supplier Thermo Scientific Chemicals Supplier No. 457490010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 603-81-6 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 603-81-6
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00137818
InChI Key KKTUQAYCCLMNOA-UHFFFAOYSA-N
Synonym diaminobenzoic acid, benzoic acid, diamino, 3-carboxy-1,2-diaminobenzene, pubchem3145, acmc-209mjn, 2,3-diaminobenzonic acid, 2,3-diamino benzoic acid, ksc203g4l, pharmabridge p-2952, 2,3-diaminobenzoic acid
PubChem CID 198069
IUPAC Name 2,3-diaminobenzoic acid
SMILES NC1=CC=CC(C(O)=O)=C1N

Specifications

Melting Point 232.0°C to 234.5°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 152.15
Percent Purity 95%
Chemical Name or Material 2,3-Diaminobenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only