accessibility menu, dialog, popup

UserName

CAS RN 603-81-6

IUPAC Name: 2,3-diaminobenzoic acid
Synonyms: 2,3-diaminobenzoic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: KKTUQAYCCLMNOA-UHFFFAOYSA-N
SMILES: NC1=CC=CC(C(O)=O)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results