missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane 98.0+%, TCI America™

Catalog No. C29081G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29081G 1 g
C29085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29081G Supplier TCI America Supplier No. C29081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3308-94-9
Molecular Formula C12H14ClFO2
Molecular Weight (g/mol) 244.69
MDL Number MFCD00020909
InChI Key FXFDJSQOCVDXBX-UHFFFAOYSA-N
Synonym 2-3-chloropropyl-2-4-fluorophenyl-1,3-dioxolane, 2-3-chloropropyl-2-p-fluorophenyl-1,3-dioxolane, pubchem2065, 4'-fluoro-4-chlorobutyrophenone ethylene ketal, gamma-chloro-p-fluorobutyrophenone ethylene ketal, 1,1-ethylenedioxy-1-4-fluorophenyl-4-chlorobutane, 4-chloro-1-4-fluorophenyl-1,1-ethylenedioxybutane, 1,3-dioxolane,2-3-chloropropyl-2-4-fluorophenyl, 2-3-chloropropyl-2-4-fluorophenyl-1,3-dioxalane, 2-4-fluorophenyl-2-3-chloropropyl-1,3-dioxolane
PubChem CID 2723778
IUPAC Name 2-(3-chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane
SMILES C1COC(O1)(CCCCl)C2=CC=C(C=C2)F

Specifications

Color Yellow
Boiling Point 112°C
Quantity 1 g
Formula Weight 244.69
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane