accessibility menu, dialog, popup

UserName

CAS RN 3308-94-9

IUPAC Name: 4-(3-chloropropyl)-4-(4-fluorophenyl)-1,3-dioxolane
Synonyms: 4-(3-chloropropyl)-4-(4-fluorophenyl)-1,3-dioxolane
Molecular Weight (g/mol): 244.69
Molecular Formula: C12H14ClFO2
InChi Key: NHKTWTJPGLFCQL-UHFFFAOYNA-N
SMILES: FC1=CC=C(C=C1)C1(CCCCl)COCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results