missing translation for 'onlineSavingsMsg'
Learn More

2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7125865
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3162200MG 200 mg
C31621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3162200MG Supplier TCI America Supplier No. C3162200MG

Chemical Identifiers

CAS 1443049-83-9
Molecular Formula C27H18ClN3
Molecular Weight (g/mol) 419.912
InChI Key MMAIMCOMSPMTKJ-UHFFFAOYSA-N
PubChem CID 90177101
IUPAC Name 2-[3-(3-chlorophenyl)phenyl]-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC=CC(=C3)C4=CC(=CC=C4)Cl)C5=CC=CC=C5

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 200 mg
Formula Weight 419.91
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3′-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine