accessibility menu, dialog, popup

UserName

NNNCl

CAS RN 1443049-83-9

IUPAC Name: 2-{3'-chloro-[1,1'-biphenyl]-3-yl}-4,6-diphenyl-1,3,5-triazine
Synonyms: 2-{3'-chloro-[1,1'-biphenyl]-3-yl}-4,6-diphenyl-1,3,5-triazine
Molecular Weight (g/mol): 419.91
Molecular Formula: C27H18ClN3
InChi Key: MMAIMCOMSPMTKJ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C1=CC(=CC=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results