missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Butanedione monoxime, For Spectrophotometric Det. of Urea, ≥99.0%, MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 31550500G
Specifications
| 75°C to 76°C; 75°C to 78°C | |
| 185°C to 186°C | |
| MFCD00002116 | |
| BDM; Biacetyl monoxime; Diacetyl monoxime | |
| ≥98.0% (N); ≥99.0% |
| 57-71-6 | |
| CH3 C(=NOH)COCH3 | |
| 500 g | |
| 101.10 |
Chemical Identifiers
| MFCD00002116 | |
| BDM; Biacetyl monoxime; Diacetyl monoxime |
Safety and Handling
RTECSNumber : EK3150000