missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione monoxime, For Spectrophotometric Det. of Urea, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187880
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014722 25 g
111014723 100 g
111014721 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111014722 Supplier Merck Emd Millipore Supplier No. 3155025G
Only null left

Chemical Identifiers

MDL Number MFCD00002116
Synonym BDM; Biacetyl monoxime; Diacetyl monoxime

Specifications

Melting Point 75°C to 76°C; 75°C to 78°C
CAS 57-71-6
Boiling Point 185°C to 186°C
Linear Formula CH3 C(=NOH)COCH3
Quantity 25 g
Formula Weight 101.10
Percent Purity ≥98.0% (N); ≥99.0%
RTECSNumber EK3150000