Learn More
2,3-Butanedione, 99%
CAS: 431-03-8 | C4H6O2 | 86.09 g/mol
Supplier: Thermo Scientific Chemicals 107651000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,3-Butanedione | |
| 431-03-8 | |
| 100.0 | |
| Yellow | |
| 88.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3910 to 1.3990 | |
| MFCD00008756 | |
| 01, 769 | |
| 0.985 | |
| 2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane | |
| QSJXEFYPDANLFS-UHFFFAOYSA-N | |
| butane-2,3-dione | |
| 650 | |
| 86.09 | |
| Liquid |
| 99% | |
| 98.5 | |
| -4°C to 2°C | |
| 0.9850g/mL | |
| 7°C | |
| 98.5% min. (GC) | |
| C4H6O2 | |
| CH3COCOCH3 | |
| 100 mL | |
| 07,21 | |
| 15, 2969 | |
| Solubility in water: 200g/L (20°C). Other solubilities: miscible with alcohol, ether, very soluble in acetone, soluble in oils, fats, propylene glycol, glycerol, soluble in carbitols, all important organic solven, ts | |
| CC(=O)C(=O)C | |
| 86.09 | |
| CHEBI:16583 | |
| 99% |
Chemical Identifiers
| 431-03-8 | |
| 86.09 | |
| QSJXEFYPDANLFS-UHFFFAOYSA-N | |
| 650 | |
| butane-2,3-dione |
| C4H6O2 | |
| MFCD00008756 | |
| 2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane | |
| CHEBI:16583 | |
| CC(=O)C(=O)C |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Toxic if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause damage to organs thro
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POIS
GHS Signal Word: Danger
EINECSNumber : 207-069-8
RTECSNumber : EK2625000
TSCA : TSCA
RUO – Research Use Only