accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 431-03-8

IUPAC Name: butane-2,3-dione
Synonyms: diacetyl dimethyl glyoxal ccris 827 diacetyl (natural) butane-2,3-dione (substance)
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: QSJXEFYPDANLFS-UHFFFAOYSA-N
SMILES: CC(=O)C(C)=O
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)