missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione, 99%

Catalog No. p-78180
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107650050 5 mL Glass bottle
AC107651000 100 mL Glass bottle
AC107655000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107650050 Supplier Thermo Scientific Chemicals Supplier No. 107650050
Only null left

CAS: 431-03-8 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 431-03-8
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008756
InChI Key QSJXEFYPDANLFS-UHFFFAOYSA-N
Synonym 2,3-butanedione, diacetyl, biacetyl, dimethylglyoxal, 2,3-diketobutane, dimethyl glyoxal, butanedione, dimethyl diketone, 2,3-butadione, 2,3-dioxobutane
PubChem CID 650
ChEBI CHEBI:16583
IUPAC Name butane-2,3-dione
SMILES CC(=O)C(=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -4°C to 2°C
Color Yellow
Density 0.9850g/mL
Boiling Point 88.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3910 to 1.3990
Linear Formula CH3COCOCH3
Quantity 5 mL
Beilstein 01, 769
Fieser 07,21
Specific Gravity 0.985
Merck Index 15, 2969
Solubility Information Solubility in water: 200g/L (20°C). Other solubilities: miscible with alcohol, ether, very soluble in acetone, soluble in oils, fats, propylene glycol, glycerol, soluble in carbitols, all important organic solven, ts
Formula Weight 86.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2,3-Butanedione, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Toxic if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause damage to organs thro
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POIS
EINECSNumber 207-069-8
RTECSNumber EK2625000
TSCA TSCA

RUO – Research Use Only