missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)triphenylene 98.0+%, TCI America™

Catalog No. B4746200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4746200MG 200 mg
B47461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4746200MG Supplier TCI America Supplier No. B4746200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1313514-53-2
Molecular Formula C24H15Br
Molecular Weight (g/mol) 383.288
InChI Key KWXFBIBEVROWEF-UHFFFAOYSA-N
PubChem CID 58472032
IUPAC Name 2-(3-bromophenyl)triphenylene
SMILES C1=CC=C2C(=C1)C3=C(C=C(C=C3)C4=CC(=CC=C4)Br)C5=CC=CC=C25

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 200 mg
Formula Weight 383.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Bromophenyl)triphenylene