accessibility menu, dialog, popup

UserName

CAS RN 1313514-53-2

IUPAC Name: 2-(3-bromophenyl)triphenylene
Synonyms: 2-(3-bromophenyl)triphenylene
Molecular Weight (g/mol): 383.29
Molecular Formula: C24H15Br
InChi Key: KWXFBIBEVROWEF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results