missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)naphthalene 95.0+%, TCI America™

Catalog No. B45681G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45681G 1 g
B45685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45681G Supplier TCI America Supplier No. B45681G

Chemical Identifiers

CAS 667940-23-0
Molecular Formula C16H11Br
Molecular Weight (g/mol) 283.17
MDL Number MFCD16658911
InChI Key FWPXWVYUNHYGPE-UHFFFAOYSA-N
Synonym 2-3-bromophenyl naphthalene, naphthalene, 2-3-bromophenyl, 2 3-bromophenyl naphthalene, ksc279g1j
PubChem CID 23080773
IUPAC Name 2-(3-bromophenyl)naphthalene
SMILES BrC1=CC=CC(=C1)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 1 g
Formula Weight 283.17
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Bromophenyl)naphthalene