accessibility menu, dialog, popup

UserName

Br

CAS RN 667940-23-0

IUPAC Name: 2-(3-bromophenyl)naphthalene
Synonyms: 2-(3-bromophenyl)naphthalene
Molecular Weight (g/mol): 283.17
Molecular Formula: C16H11Br
InChi Key: FWPXWVYUNHYGPE-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=CC2=CC=CC=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results