missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine 98.0+%, TCI America™

Catalog No. B36545G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36545G 5 g
B36541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36545G Supplier TCI America Supplier No. B36545G

Chemical Identifiers

CAS 927384-43-8
Molecular Formula C16H12BBrN2
Molecular Weight (g/mol) 323
MDL Number MFCD16038145
InChI Key RDGOAMRKYXEHHS-UHFFFAOYSA-N
Synonym 3-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
PubChem CID 53384412
SMILES B1(NC2=CC=CC3=C2C(=CC=C3)N1)C4=CC(=CC=C4)Br

Specifications

Quantity 5 g
Formula Weight 323.00
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine
TSCA No
Recommended Storage Refrigerator