CAS RN 927384-43-8
IUPAC Name:
3-(3-bromophenyl)-2,4-diaza-3-boratricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
Synonyms:
3-(3-bromophenyl)-2,4-diaza-3-boratricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
Molecular Weight (g/mol):
323
Molecular Formula:
C16H12BBrN2
InChi Key:
RDGOAMRKYXEHHS-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=C1)B1NC2=CC=CC3=CC=CC(N1)=C23
1
–
1
of
1
results
1
2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine 98.0+%, TCI America™
CAS: 927384-43-8 Molecular Formula: C16H12BBrN2 Molecular Weight (g/mol): 323 MDL Number: MFCD16038145 InChI Key: RDGOAMRKYXEHHS-UHFFFAOYSA-N Synonym: 3-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine PubChem CID: 53384412 SMILES: B1(NC2=CC=CC3=C2C(=CC=C3)N1)C4=CC(=CC=C4)Br