missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(4-methoxyphenyl)-5-(4-cyanophenyl)tetrazolium Chloride 90.0+%, TCI America™

Catalog No. b1311100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1311100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1311100MG Supplier TCI America Supplier No. B1311100MG

Chemical Identifiers

CAS 151390-91-9
Molecular Formula C22H18ClN5O2
Molecular Weight (g/mol) 419.869
MDL Number MFCD00060131
InChI Key FSMGZHHFVGWWGM-UHFFFAOYSA-M
PubChem CID 44630129
IUPAC Name 4-[2,3-bis(4-methoxyphenyl)tetrazol-2-ium-5-yl]benzonitrile;chloride
SMILES COC1=CC=C(C=C1)N2N=C(N=[N+]2C3=CC=C(C=C3)OC)C4=CC=C(C=C4)C#N.[Cl-]

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 419.87
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(4-methoxyphenyl)-5-(4-cyanophenyl)tetrazolium Chloride