CAS RN 151390-91-9
IUPAC Name:
5-(4-cyanophenyl)-2,3-bis(4-methoxyphenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms:
5-(4-cyanophenyl)-2,3-bis(4-methoxyphenyl)-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol):
419.87
Molecular Formula:
C22H18ClN5O2
InChi Key:
FSMGZHHFVGWWGM-UHFFFAOYSA-M
SMILES:
[Cl-].COC1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=C(OC)C=C1)C1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
2,3-Bis(4-methoxyphenyl)-5-(4-cyanophenyl)tetrazolium Chloride 90.0+%, TCI America™
CAS: 151390-91-9 Molecular Formula: C22H18ClN5O2 Molecular Weight (g/mol): 419.869 MDL Number: MFCD00060131 InChI Key: FSMGZHHFVGWWGM-UHFFFAOYSA-M PubChem CID: 44630129 IUPAC Name: 4-[2,3-bis(4-methoxyphenyl)tetrazol-2-ium-5-yl]benzonitrile;chloride SMILES: COC1=CC=C(C=C1)N2N=C(N=[N+]2C3=CC=C(C=C3)OC)C4=CC=C(C=C4)C#N.[Cl-]