missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trimethylphenol, 95%

CAS: 2416-94-6 | C9H12O | 136.19 g/mol

Supplier:  Thermo Scientific Chemicals H6048922

 View more versions of this product

Catalog No. AAH6048922


Only null left
Explore available promotions

Description

Description

2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Specifications

Specifications

2,3,6-Trimethylphenol
59°C to 62°C
215°C
C9H12O
100 g
2042207
Soluble in methanol.
CC1=CC=C(C)C(O)=C1C
136.19
136.19
2416-94-6
0.94
100°C (212°F)
MFCD00002229
UN2430
3-hydroxypseudocumene, phenol, 2,3,6-trimethyl, 1-hydroxy-2,3,6-trimethylbenzene, 2,3,6-trimethyl-phenol, unii-05wkl2l5lj, 2,3,6-trimethyl phenol, 05wkl2l5lj, dsstox_cid_2187, dsstox_rid_76518, dsstox_gsid_22187
QQOMQLYQAXGHSU-UHFFFAOYSA-N
2,3,6-trimethylphenol
17016
95%

Chemical Identifiers

2416-94-6
136.19
QQOMQLYQAXGHSU-UHFFFAOYSA-N
17016
CC1=CC=C(C)C(O)=C1C
C9H12O
MFCD00002229
3-hydroxypseudocumene, phenol, 2,3,6-trimethyl, 1-hydroxy-2,3,6-trimethylbenzene, 2,3,6-trimethyl-phenol, unii-05wkl2l5lj, 2,3,6-trimethyl phenol, 05wkl2l5lj, dsstox_cid_2187, dsstox_rid_76518, dsstox_gsid_22187
2,3,6-trimethylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c

H315-H317-H318

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.

EINECSNumber : 219-330-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only