missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trimethylphenol, 95%

Catalog No. p-7058104
Change view
Click to view available options
Quantity:
100 g
25 g
Catalog No. Quantity
AAH6048914 25 g
AAH6048922 100 g
2 options

Catalog No. AAH6048914

Supplier: Thermo Scientific Chemicals H6048914

Only null left

CAS: 2416-94-6 | C9H12O | 136.19 g/mol

2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trimethylphenol is used as intermediate for synthetic vitamin E. It is used for manufacturing 2, 3, 5-Trimethylhydroquinone. It is used as intermediate for antioxidants and plastics. It is used as a comonomer for the modification of polyphenylene oxide resins.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.

Chemical Identifiers

CAS 2416-94-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002229
InChI Key QQOMQLYQAXGHSU-UHFFFAOYSA-N
Synonym 3-hydroxypseudocumene, phenol, 2,3,6-trimethyl, 1-hydroxy-2,3,6-trimethylbenzene, 2,3,6-trimethyl-phenol, unii-05wkl2l5lj, 2,3,6-trimethyl phenol, 05wkl2l5lj, dsstox_cid_2187, dsstox_rid_76518, dsstox_gsid_22187
PubChem CID 17016
IUPAC Name 2,3,6-trimethylphenol
SMILES CC1=CC=C(C)C(O)=C1C

Specifications

Melting Point 59°C to 62°C
Density 0.94
Boiling Point 215°C
Flash Point 100°C (212°F)
Quantity 25 g
UN Number UN2430
Beilstein 2042207
Solubility Information Soluble in methanol.
Formula Weight 136.19
Percent Purity 95%
Chemical Name or Material 2,3,6-Trimethylphenol
Show More Show Less
Hazard Category H315-H317-H318
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber 219-330-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only