Learn More
2,3,5-Trimethylpyrazine, 99%
CAS: 14667-55-1 | C7H10N2 | 122.171 g/mol
$69.56 - $866.77
Chemical Identifiers
| CAS | 14667-55-1 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.171 |
| MDL Number | MFCD00006145 |
| InChI Key | IAEGWXHKWJGQAZ-UHFFFAOYSA-N |
| Synonym | trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl |
| PubChem CID | 26808 |
| IUPAC Name | 2,3,5-trimethylpyrazine |
| SMILES | CC1=CN=C(C(=N1)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1487009
|
Thermo Scientific Chemicals
A1487009 |
10 g |
Each for $69.56
|
|
|||||
|
AAA1487018
|
Thermo Scientific Chemicals
A1487018 |
50 g |
Each for $206.53
|
|
|||||
|
AAA1487030
|
Thermo Scientific Chemicals
A1487030 |
250 g |
Each for $866.77
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14667-55-1 | |
| 122.171 | |
| IAEGWXHKWJGQAZ-UHFFFAOYSA-N | |
| 26808 | |
| CC1=CN=C(C(=N1)C)C |
| C7H10N2 | |
| MFCD00006145 | |
| trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl | |
| 2,3,5-trimethylpyrazine |
Specifications
| 14667-55-1 | |
| 171°C to 172°C | |
| C7H10N2 | |
| MFCD00006145 | |
| UN1993 | |
| trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl | |
| CC1=CN=C(C(=N1)C)C | |
| 122.171 | |
| 122.17 | |
| 2,3,5-Trimethylpyrazine |
| 0.979 | |
| 54°C (129°F) | |
| 1.505 | |
| 10 g | |
| 2423 | |
| IAEGWXHKWJGQAZ-UHFFFAOYSA-N | |
| 2,3,5-trimethylpyrazine | |
| 26808 | |
| 99% |
Safety and Handling
GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 238-712-0
RTECSNumber : UQ3907000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only