missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylpyrazine, 99%

CAS: 14667-55-1 | C7H10N2 | 122.171 g/mol

Supplier:  Thermo Scientific Chemicals A1487018

 View more versions of this product

Catalog No. AAA1487018

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,3,5-Trimethylpyrazine
0.979
54°C (129°F)
1.505
50 g
2423
IAEGWXHKWJGQAZ-UHFFFAOYSA-N
2,3,5-trimethylpyrazine
26808
99%
14667-55-1
171°C to 172°C
C7H10N2
MFCD00006145
UN1993
trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl
CC1=CN=C(C(=N1)C)C
122.171
122.17

Chemical Identifiers

14667-55-1
122.171
IAEGWXHKWJGQAZ-UHFFFAOYSA-N
26808
CC1=CN=C(C(=N1)C)C
C7H10N2
MFCD00006145
trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl
2,3,5-trimethylpyrazine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c

H226-H302

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 238-712-0

RTECSNumber : UQ3907000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only