accessibility menu, dialog, popup

UserName

CH3NCH3H3CN

CAS RN 14667-55-1

IUPAC Name: 2,3,5-trimethylpyrazine
Synonyms: trimethylpyrazine pyrazine, trimethyl-
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: IAEGWXHKWJGQAZ-UHFFFAOYSA-N
SMILES: CC1=CN=C(C)C(C)=N1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)