missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Trimethylanisole 97.0+%, TCI America™
$106.25 - $106.25
Chemical Identifiers
| CAS | 20469-61-8 |
|---|---|
| Molecular Formula | C10H14O |
| Molecular Weight (g/mol) | 150.22 |
| MDL Number | MFCD00142783 |
| InChI Key | AWONIZVBKXHWJP-UHFFFAOYSA-N |
| Synonym | 2,3,5-trimethylanisole, anisole, 2,3,5-trimethyl, benzene, 1-methoxy-2,3,5-trimethyl, 1-methoxy-2,3,5-trimethyl-benzene, pubchem2688, acmc-209fam, 2,3,5-trimethylanizole, 3-methoxy-1,2,5-trimethylbenzene |
| PubChem CID | 88555 |
| IUPAC Name | 1-methoxy-2,3,5-trimethylbenzene |
| SMILES | COC1=CC(C)=CC(C)=C1C |
Chemical Identifiers
| 20469-61-8 | |
| 150.22 | |
| AWONIZVBKXHWJP-UHFFFAOYSA-N | |
| 88555 | |
| COC1=CC(C)=CC(C)=C1C |
| C10H14O | |
| MFCD00142783 | |
| 2,3,5-trimethylanisole, anisole, 2,3,5-trimethyl, benzene, 1-methoxy-2,3,5-trimethyl, 1-methoxy-2,3,5-trimethyl-benzene, pubchem2688, acmc-209fam, 2,3,5-trimethylanizole, 3-methoxy-1,2,5-trimethylbenzene | |
| 1-methoxy-2,3,5-trimethylbenzene |
Specifications
| 20469-61-8 | |
| 216°C | |
| MFCD00142783 | |
| 2,3,5-trimethylanisole, anisole, 2,3,5-trimethyl, benzene, 1-methoxy-2,3,5-trimethyl, 1-methoxy-2,3,5-trimethyl-benzene, pubchem2688, acmc-209fam, 2,3,5-trimethylanizole, 3-methoxy-1,2,5-trimethylbenzene | |
| COC1=CC(C)=CC(C)=C1C | |
| 150.22 | |
| 150.22 | |
| Liquid |
| Yellow | |
| C10H14O | |
| 5 g | |
| AWONIZVBKXHWJP-UHFFFAOYSA-N | |
| 1-methoxy-2,3,5-trimethylbenzene | |
| 88555 | |
| ≥97.0% (GC) | |
| 2,3,5-Trimethylanisole |
Safety and Handling
TSCA : No