Learn More
2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 468640050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone | |
| 53.0°C to 55.0°C | |
| MFCD08703966 | |
| LDHBSABBBAUMCZ-UHFFFAOYNA-N | |
| 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-one | |
| 418.49 |
| 55094-52-5 | |
| C26H26O5 | |
| 5 g | |
| O=C1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1 | |
| 418.49 |
Chemical Identifiers
| 55094-52-5 | |
| 418.49 | |
| LDHBSABBBAUMCZ-UHFFFAOYNA-N | |
| O=C1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1 |
| C26H26O5 | |
| MFCD08703966 | |
| 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-one |
Safety and Handling
GHS H Statement Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO – Research Use Only