accessibility menu, dialog, popup

UserName

CAS RN 55094-52-5

IUPAC Name: 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-one
Synonyms: 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-one
Molecular Weight (g/mol): 418.49
Molecular Formula: C26H26O5
InChi Key: LDHBSABBBAUMCZ-UHFFFAOYNA-N
SMILES: O=C1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results