missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone, Thermo Scientific Chemicals

Catalog No. p-7214835
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468640010 1 g
AC468640050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468640010 Supplier Thermo Scientific Chemicals Supplier No. 468640010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55094-52-5
Molecular Formula C26H26O5
Molecular Weight (g/mol) 418.49
MDL Number MFCD08703966
InChI Key LDHBSABBBAUMCZ-UHFFFAOYNA-N
IUPAC Name 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-one
SMILES O=C1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1

Specifications

Melting Point 53.0°C to 55.0°C
Quantity 1 g
Formula Weight 418.49
Chemical Name or Material 2,3,5-Tri-O-benzyl-D-ribono-1,4-lactone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Harmful in contact with skin.

Harmful if inhaled.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only