missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorothiophenol, 98%

CAS: 769-40-4 | C6H2F4S | 182.136 g/mol

$201.25 - $676.11

Chemical Identifiers

CAS 769-40-4
Molecular Formula C6H2F4S
Molecular Weight (g/mol) 182.136
MDL Number MFCD00004829
InChI Key IGOGJHYWSOZGAE-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluorothiophenol, 2,3,5,6-tetrafluorobenzene-1-thiol, benzenethiol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorbenzolthiol, timtec-bb sbb000230, #, 2,3,5,6-tetrafluorophenyl hydrosulfide, 2,3,5,6-tetrafluorobenzenethiol
PubChem CID 69859
IUPAC Name 2,3,5,6-tetrafluorobenzenethiol
SMILES C1=C(C(=C(C(=C1F)F)S)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2348306
View Documents
Thermo Scientific Chemicals
B2348306
5 g
Each for $201.25
Only null left
 
AAB2348314
View Documents
Thermo Scientific Chemicals
B2348314
25 g
Each for $676.11
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

769-40-4
182.136
IGOGJHYWSOZGAE-UHFFFAOYSA-N
69859
C1=C(C(=C(C(=C1F)F)S)F)F
C6H2F4S
MFCD00004829
2,3,5,6-tetrafluorothiophenol, 2,3,5,6-tetrafluorobenzene-1-thiol, benzenethiol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorbenzolthiol, timtec-bb sbb000230, #, 2,3,5,6-tetrafluorophenyl hydrosulfide, 2,3,5,6-tetrafluorobenzenethiol
2,3,5,6-tetrafluorobenzenethiol

Specifications

769-40-4
152°C to 153°C
Stench
1.485
5 g
Air Sensitive
IGOGJHYWSOZGAE-UHFFFAOYSA-N
2,3,5,6-tetrafluorobenzenethiol
69859
98%
1.53
48°C (119°F)
C6H2F4S
MFCD00004829
UN1992
2,3,5,6-tetrafluorothiophenol, 2,3,5,6-tetrafluorobenzene-1-thiol, benzenethiol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorbenzolthiol, timtec-bb sbb000230, #, 2,3,5,6-tetrafluorophenyl hydrosulfide, 2,3,5,6-tetrafluorobenzenethiol
C1=C(C(=C(C(=C1F)F)S)F)F
182.136
182.14
2,3,5,6-Tetrafluorothiophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 212-210-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only