missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorobenzyl Alcohol 96.0+%, TCI America™

Catalog No. T18841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18841G 1 g
T18845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18841G Supplier TCI America Supplier No. T18841G

Chemical Identifiers

CAS 4084-38-2
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.10
MDL Number MFCD00792428
InChI Key AGWVQASYTKCTCC-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluorobenzyl alcohol, 2,3,5,6-tetrafluorophenyl methanol, 2,3,5,6-tetrafluorobenzylalcohol, benzenemethanol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorophenyl methan-1-ol, 2,3,5,6-tetrafluorobenyl alcohol, pubchem15529, acmc-209jh7, 2,3,5,6-tetrafluorbenzyl alcohol, benzenemethanol,2,3,5,6-tetrafluoro
PubChem CID 2734029
IUPAC Name (2,3,5,6-tetrafluorophenyl)methanol
SMILES OCC1=C(F)C(F)=CC(F)=C1F

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 180.10
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3,5,6-Tetrafluorobenzyl Alcohol
EINECSNumber (3)-4558
TSCA No