missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7137001
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18811G 1 g
T18815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18811G Supplier TCI America Supplier No. T18811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 652-18-6
Molecular Formula C7H2F4O2
Molecular Weight (g/mol) 194.085
MDL Number MFCD00002408
InChI Key KVLBXIOFJUWSJQ-UHFFFAOYSA-N
Synonym benzoic acid, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorobenzoicacid, pubchem1341, acmc-1avk1, benzoic acid,2,3,5,6-tetrafluoro
PubChem CID 567533
IUPAC Name 2,3,5,6-tetrafluorobenzoic acid
SMILES C1=C(C(=C(C(=C1F)F)C(=O)O)F)F

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 1 g
Formula Weight 194.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrafluorobenzoic Acid