CAS RN 652-18-6
IUPAC Name:
2,3,5,6-tetrafluorobenzoate
Synonyms:
2,3,5,6-tetrafluorobenzoate
Molecular Weight (g/mol):
193.08
Molecular Formula:
C7HF4O2
InChi Key:
KVLBXIOFJUWSJQ-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=C(F)C(F)=CC(F)=C1F
1
–
1
of
1
results
1
2,3,5,6-Tetrafluorobenzoic Acid 98.0+%, TCI America™
CAS: 652-18-6 Molecular Formula: C7H2F4O2 Molecular Weight (g/mol): 194.085 MDL Number: MFCD00002408 InChI Key: KVLBXIOFJUWSJQ-UHFFFAOYSA-N Synonym: benzoic acid, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluorobenzoicacid,pubchem1341,acmc-1avk1,benzoic acid,2,3,5,6-tetrafluoro PubChem CID: 567533 IUPAC Name: 2,3,5,6-tetrafluorobenzoic acid SMILES: C1=C(C(=C(C(=C1F)F)C(=O)O)F)F