missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorobenzaldehyde 94.0+%, TCI America™

Catalog No. T18985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18985G 5 g
T18981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18985G Supplier TCI America Supplier No. T18985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19842-76-3
Molecular Formula C7H2F4O
Molecular Weight (g/mol) 178.086
MDL Number MFCD00167387
InChI Key YIRYOMXPMOLQSO-UHFFFAOYSA-N
Synonym benzaldehyde,2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-benzaldehyde, pubchem10080, 1,2,4,5-tetrafluorobenzaldehyde, 2,3,5,6-tetrafluorobenzaldehyde, benzaldehyde, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrakis fluoranyl benzaldehyde
PubChem CID 601331
IUPAC Name 2,3,5,6-tetrafluorobenzaldehyde
SMILES C1=C(C(=C(C(=C1F)F)C=O)F)F

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
Formula Weight 178.09
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5,6-Tetrafluorobenzaldehyde