accessibility menu, dialog, popup

UserName

CAS RN 19842-76-3

FFFOF

CAS RN 19842-76-3

IUPAC Name: 2,3,5,6-tetrafluorobenzaldehyde
Synonyms: 2,3,5,6-tetrafluorobenzaldehyde
Molecular Weight (g/mol): 178.09
Molecular Formula: C7H2F4O
InChi Key: YIRYOMXPMOLQSO-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(C=O)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results