missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoro-p-xylene 98.0+%, TCI America™

Catalog No. T19471G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19471G 1 g
T19475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19471G Supplier TCI America Supplier No. T19471G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 703-87-7
Molecular Formula C8H6F4
Molecular Weight (g/mol) 178.13
MDL Number MFCD00012231
InChI Key IWKPBYPUIPVYNZ-UHFFFAOYSA-N
Synonym 2,3,5,6-Tetrafluoro-1,4-dimethylbenzene
PubChem CID 136549
IUPAC Name 1,2,4,5-tetrafluoro-3,6-dimethylbenzene
SMILES CC1=C(C(=C(C(=C1F)F)C)F)F

Specifications

Melting Point 35°C
Color White
Quantity 1 g
Physical Form Crystal-Powder at 20°C
UN Number 1325
Formula Weight 178.13
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,3,5,6-Tetrafluoro-p-xylene