accessibility menu, dialog, popup

UserName

CAS RN 703-87-7

IUPAC Name: 1,2,4,5-tetrafluoro-3,6-dimethylbenzene
Synonyms: 2,3,5,6-tetrafluoro-p-xylene
Molecular Weight (g/mol): 178.13
Molecular Formula: C8H6F4
InChi Key: IWKPBYPUIPVYNZ-UHFFFAOYSA-N
SMILES: CC1=C(F)C(F)=C(C)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results