missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 95%

CAS: 29261-33-4 | C12F4N4 | 276.16 g/mol

Supplier:  Thermo Scientific Chemicals 312670050

Catalog No. AC312670050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane
291.0°C
Glass bottle
5 mg
Solubility in water: insoluble. Other solubilities: soluble in chloroform, dichloromethane,, acetic acid
C(#N)C(=C1C(=C(C(=C(C#N)C#N)C(=C1F)F)F)F)C#N
276.16
276.16
29261-33-4
94% min
C12F4N4
2,3,5,6-tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 2,2'-perfluorocyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tcnqf4, 2,3,5,6-tetrafluorotetracyanoquinodimethane, f4-tcnq, tetrafluorotetracyanoquinodimethane, 2-4-dicyanomethylidene-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene propanedinitrile, f4tcnq, tetrafluorotetracyanoquinodimethane anion radical, tetrafluorotetracyanoquinodimethane purifiedbysubl
IXHWGNYCZPISET-UHFFFAOYSA-N
2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
2733307
95%

Chemical Identifiers

29261-33-4
276.16
2,3,5,6-tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 2,2'-perfluorocyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tcnqf4, 2,3,5,6-tetrafluorotetracyanoquinodimethane, f4-tcnq, tetrafluorotetracyanoquinodimethane, 2-4-dicyanomethylidene-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene propanedinitrile, f4tcnq, tetrafluorotetracyanoquinodimethane anion radical, tetrafluorotetracyanoquinodimethane purifiedbysubl
2-[4-(dicyanomethylidene)-2,3,5,6-tetrafluorocyclohexa-2,5-dien-1-ylidene]propanedinitrile
C12F4N4
IXHWGNYCZPISET-UHFFFAOYSA-N
2733307
C(#N)C(=C1C(=C(C(=C(C#N)C#N)C(=C1F)F)F)F)C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only