missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoro-4-methylbenzoic Acid 97.0+%, TCI America™

Catalog No. T23231G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23231G Supplier TCI America Supplier No. T23231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 652-32-4
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
InChI Key COOULIOYEXBFDT-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-p-toluic acid, 4-methyl-2,3,5,6-tetrafluorobenzoic acid, p-toluic acid, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-4-methyl-benzoic acid, tetrafluoro-p-toluic acid, acmc-1az53, 2,3,5,6-tetrafluoro-4-methylbenzoicacid, 2,3,5,6-tetrafluoro-4-toluic acid, 4-carboxy-2,3,5,6-tetrafluorotoluene
PubChem CID 69548
IUPAC Name 2,3,5,6-tetrafluoro-4-methylbenzoic acid
SMILES CC1=C(C(=C(C(=C1F)F)C(=O)O)F)F

Specifications

Melting Point 171°C
Quantity 1 g
Formula Weight 208.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrafluoro-4-methylbenzoic Acid