accessibility menu, dialog, popup

UserName

CAS RN 652-32-4

IUPAC Name: 2,3,5,6-tetrafluoro-4-methylbenzoic acid
Synonyms: 2,3,5,6-tetrafluoro-4-methylbenzoic acid
Molecular Weight (g/mol): 208.11
Molecular Formula: C8H4F4O2
InChi Key: COOULIOYEXBFDT-UHFFFAOYSA-N
SMILES: CC1=C(F)C(F)=C(C(O)=O)C(F)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results