missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluoro-4-mercaptobenzoic Acid 98.0+%, TCI America™

Catalog No. T25421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25421G 1 g
T25425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25421G Supplier TCI America Supplier No. T25421G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5211-44-9
Molecular Formula C7H2F4O2S
Molecular Weight (g/mol) 226.14
MDL Number MFCD00129954
InChI Key USFMEWZQIHKRDP-UHFFFAOYSA-N
PubChem CID 297543
IUPAC Name 2,3,5,6-tetrafluoro-4-sulfanylbenzoic acid
SMILES OC(=O)C1=C(F)C(F)=C(S)C(F)=C1F

Specifications

Melting Point 153°C
Quantity 1 g
Formula Weight 226.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrafluoro-4-mercaptobenzoic Acid