missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5,6-Tetrafluoro-4-hydroxybenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 154°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 210.08 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,3,5,6-Tetrafluoro-4-hydroxybenzoic Acid |
Safety and Handling
| TSCA | No |