CAS RN 652-34-6
IUPAC Name:
2,3,5,6-tetrafluoro-4-hydroxybenzoic acid
Synonyms:
2,3,5,6-tetrafluoro-4-hydroxybenzoic acid
Molecular Weight (g/mol):
210.08
Molecular Formula:
C7H2F4O3
InChi Key:
FTLHGQOBAPTEHE-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(F)C(F)=C(O)C(F)=C1F
1
–
1
of
1
results
1
2,3,5,6-Tetrafluoro-4-hydroxybenzoic Acid 98.0+%, TCI America™
CAS: 652-34-6 Molecular Formula: C7H2F4O3 Molecular Weight (g/mol): 210.084 MDL Number: MFCD00129952 InChI Key: FTLHGQOBAPTEHE-UHFFFAOYSA-N Synonym: 4-hydroxy-2,3,5,6-tetrafluorobenzoic acid,2,3,5,6-tetrafluoro-4-hydroxy-benzoic acid,tetrafluoro-4-hydroxybenzoic acid,benzoic acid, 2,3,5,6-tetrafluoro-4-hydroxy,f4-pohb,tetrafluoro-4-hydroxybenzoate,zlchem 412,acmc-1b1y5,ksc622i2b,4-carboxy-2,3,5,6-tetrafluorophenol PubChem CID: 98835 IUPAC Name: 2,3,5,6-tetrafluoro-4-hydroxybenzoic acid SMILES: C1(=C(C(=C(C(=C1F)F)O)F)F)C(=O)O