missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzoic Acid Hydrate 98.0+%, TCI America™

Catalog No. P05711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P05711G 1 g
P05715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P05711G Supplier TCI America Supplier No. P05711G
Only null left

Chemical Identifiers

CAS 610-02-6
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002447
InChI Key BRRSNXCXLSVPFC-UHFFFAOYSA-N
Synonym 4-pyrogallolcarboxylic acid, pyrogallolcarboxylic acid, 2,3,4-trihydroxybenzoate, unii-ad1id2jf5o, benzoic acid, trihydroxy, benzoic acid, 2,3,4-trihydroxy, pyrogallol-4-carboxylic acid, 2,3,4-trihydroxybenzene carboxylic acid, ad1id2jf5o, pyrogallolcarboxylate
PubChem CID 11874
IUPAC Name 2,3,4-trihydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C(O)=C1O

Specifications

Quantity 1 g
Formula Weight 170.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4-Trihydroxybenzoic Acid Hydrate
RTECSNumber DH8900000
TSCA No