accessibility menu, dialog, popup

UserName

HOOOHHOHO

CAS RN 610-02-6

IUPAC Name: 2,3,4-trihydroxybenzoic acid
Synonyms: 2,3,4-trihydroxybenzoic acid
Molecular Weight (g/mol): 170.12
Molecular Formula: C7H6O5
InChi Key: BRRSNXCXLSVPFC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C(O)=C1O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)