missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzoic acid, 97%

Catalog No. AAB2037114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2037114 25 g
AAB2037103 1 g
AAB2037106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2037114 Supplier Thermo Scientific Chemicals Supplier No. B2037114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 610-02-6 | C7H6O5 | 170.12 g/mol

2,3,4-Trihydroxybenzoic acid is used as a reagent during catalytic spectrophotometric determination of vanadium in natural and sea waters and used in the pharmaceuticals. It is a internal standard in separation of phenolic acids by HPLC with photodiode array detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4-Trihydroxybenzoic acid is used as a reagent during catalytic spectrophotometric determination of vanadium in natural and sea waters and used in the pharmaceuticals. It is a internal standard in separation of phenolic acids by HPLC with photodiode array detection.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 610-02-6
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002447
InChI Key BRRSNXCXLSVPFC-UHFFFAOYSA-N
Synonym 4-pyrogallolcarboxylic acid, pyrogallolcarboxylic acid, 2,3,4-trihydroxybenzoate, unii-ad1id2jf5o, benzoic acid, trihydroxy, benzoic acid, 2,3,4-trihydroxy, pyrogallol-4-carboxylic acid, 2,3,4-trihydroxybenzene carboxylic acid, ad1id2jf5o, pyrogallolcarboxylate
PubChem CID 11874
IUPAC Name 2,3,4-trihydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C(O)=C1O

Specifications

Melting Point ∼210°C (decomposition)
Quantity 25 g
Beilstein 2210807
Solubility Information Slightly soluble in water.
Formula Weight 170.12
Percent Purity 97%
Chemical Name or Material 2,3,4-Trihydroxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-205-9
RTECSNumber DH8900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only