missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trifluorotoluene 97.0+%, TCI America™
$140.54 - $456.26
Chemical Identifiers
| CAS | 193533-92-5 |
|---|---|
| Molecular Formula | C7H5F3 |
| Molecular Weight (g/mol) | 146.112 |
| MDL Number | MFCD00075194 |
| InChI Key | LRQPEHJWTXCLQY-UHFFFAOYSA-N |
| PubChem CID | 2777991 |
| IUPAC Name | 1,2,3-trifluoro-4-methylbenzene |
| SMILES | CC1=C(C(=C(C=C1)F)F)F |
Chemical Identifiers
| 193533-92-5 | |
| 146.112 | |
| LRQPEHJWTXCLQY-UHFFFAOYSA-N | |
| 1,2,3-trifluoro-4-methylbenzene |
| C7H5F3 | |
| MFCD00075194 | |
| 2777991 | |
| CC1=C(C(=C(C=C1)F)F)F |
Specifications
| 193533-92-5 | |
| 127°C | |
| MFCD00075194 | |
| 1993 | |
| LRQPEHJWTXCLQY-UHFFFAOYSA-N | |
| 1,2,3-trifluoro-4-methylbenzene | |
| 2777991 | |
| ≥97.0% (GC) | |
| 2,3,4-Trifluorotoluene |
| Yellow | |
| C7H5F3 | |
| 1 g | |
| 2,3,4-trifluorotoluene, benzene, 1,2,3-trifluoro-4-methyl, triflurotoluene, 1,2,3-trifluoro-4-methyl-benzene, pubchem1628, acmc-1cn3t, ksc541e8d, benzene,1,2,3-trifluoro-4-methyl, benzene, 1,2,3-trifluoro-4-methyl-9ci | |
| CC1=C(C(=C(C=C1)F)F)F | |
| 146.112 | |
| 146.11 | |
| Liquid |
Safety and Handling
TSCA : No