CAS RN 193533-92-5
CAS RN 193533-92-5
IUPAC Name:
1,2,3-trifluoro-4-methylbenzene
Synonyms:
1,2,3-trifluoro-4-methylbenzene
Molecular Weight (g/mol):
146.11
Molecular Formula:
C7H5F3
InChi Key:
LRQPEHJWTXCLQY-UHFFFAOYSA-N
SMILES:
CC1=CC=C(F)C(F)=C1F
1
–
1
of
1
results
1
2,3,4-Trifluorotoluene 97.0+%, TCI America™
CAS: 193533-92-5 Molecular Formula: C7H5F3 Molecular Weight (g/mol): 146.112 MDL Number: MFCD00075194 InChI Key: LRQPEHJWTXCLQY-UHFFFAOYSA-N PubChem CID: 2777991 IUPAC Name: 1,2,3-trifluoro-4-methylbenzene SMILES: CC1=C(C(=C(C=C1)F)F)F